7-Chloro-1,2,3,4-tetrahydroisoquinoline
Catalog # APOSOR015043-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.636 g/mol
Storage Temperature:Ambient
CAS Number:82771-60-6
Formula:C₉H₁₀ClN
MDL Number:MFCD05861543