

2-Chlorostyrene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2039-87-4
:
Formula:C₈H₇Cl
MDL Number:MFCD00000567
:138.6 g/mol
Boiling Pt:58…60 °C/7mmHg
Melting Pt:–63 °C
Density:1.08 g/cm³ (25 °C)
Flash Pt:58 °C
Storage Temperature:Refrigerator
EINECS:218-026-8
UN:1993
ADR:3,III
:GHS02
:GHS07