Order Entry
Germany
ContactUsLinkComponent
Strontium carbonate 99.99%
Strontium carbonate 99.99%
Catalog # APOSIN3346-25G
Supplier:  Apollo Scientific
CAS Number:  
Strontium carbonate 99.99%
Catalog # APOSIN3346-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:1497 °C
Density:3.7 g/cm³ (25 °C)
 :147.63 g/mol
EINECS:216-643-7
 :GHS07
Storage Temperature:Ambient
CAS Number:1633-05-2
Formula:CO₃Sr
MDL Number:MFCD00011250