0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Sodium bromide
Sodium bromide, anhydrous 99.999%
Sodium bromide, anhydrous 99.999%
:
Apollo Scientific
Sodium bromide, anhydrous 99.999%
:
Apollo Scientific
:
7647-15-6
:
:
NaBr
:
MFCD00003475
:
102.89 g/mol
:
1390 °C
:
755 °C
:
3.21 g/cm³ (25 °C)
:
Ambient