Order Entry
Germany
ContactUsLinkComponent
Potassium sorbate 98%
Potassium sorbate 98%
Catalog # APOSIN2901-1KG
Supplier:  Apollo Scientific
CAS Number:  
Potassium sorbate 98%
Catalog # APOSIN2901-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :150.22 g/mol
Melting Pt:210 °C (dec.)
Density:1.36 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:24634-61-5
 :590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546