Lead ACS
Bottle
Catalog # APOSIN2182-100G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Boiling Pt:1740 °C
- Melting Pt:327.43 °C
- Density:11.34 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



