

Iron(II) chloride tetrahydrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13478-10-9
:
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709
Melting Pt:105…110 °C
Density:1.93 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-843-4
UN:3260
ADR:8,II
:198.81 g/mol
:GHS05
:GHS07
Frequently Bought Together