
Gallium(III) chloride, anhydrous 99.999%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13450-90-3
:
Formula:Cl₃Ga
MDL Number:MFCD00011018
:176.08 g/mol
Melting Pt:78 °C
Density:2.47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:236-610-0
UN:3260
ADR:8,II
:GHS05