
Ammonium carbonate 99.99%
Catalog # APOSIN1051-250G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:96.09 g/mol
Melting Pt:58 °C
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:8000-73-5
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890