2,3,5-Triphenyltetrazolium chloride (TTC)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:298-96-4
:
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963
Melting Pt:249…251 °C
:334.81 g/mol
EINECS:206-071-6
:GHS07
Storage Temperature:Refrigerator