Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(-)-Tryptophan
L(-)-Tryptophan ≥99.5%
L(-)-Tryptophan ≥99.5%
L(-)-Tryptophan ≥99.5%
Supplier:
Apollo Scientific
Certificates
Expand image
L(-)-Tryptophan ≥99.5%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
73-22-3
:
Formula:
C₁₁H₁₂N₂O₂
MDL Number:
MFCD00064340
:
204.23 g/mol
Melting Pt:
280…285 °C (dec.)
EINECS:
200-795-6
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation