Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetracycline hydrochloride
Tetracycline hydrochloride
Tetracycline hydrochloride
Tetracycline hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Tetracycline hydrochloride
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
64-75-5
:
Formula:
C₂₂H₂₅ClN₂O₈
MDL Number:
MFCD00078142
:
480.9 g/mol
Melting Pt:
220…223 °C
Storage Temperature:
Freezer
Related Information
Documentation
Certificates
Search by Lot Number
Documentation