Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Supplier:
Apollo Scientific
Certificates
Expand image
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Supplier:
Apollo Scientific
About this item
CAS Number:
480-19-3
:
Formula:
C₁₆H₁₂O₇
MDL Number:
MFCD00017310
:
316.27 g/mol
Storage Temperature:
Ambient