Order Entry
Germany
ContactUsLinkComponent
MK-1064 97%
MK-1064 97%
Catalog # APOSBISN0036-1MG
Supplier:  Apollo Scientific
CAS Number:  
MK-1064 97%
Catalog # APOSBISN0036-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :461.9 g/mol
Storage Temperature:Refrigerator
CAS Number:1207253-08-4
Formula:C₂₄H₂₀ClN₅O₃
MDL Number:MFCD28502027