Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
t-Boc-N-amido-PEG4-alcohol
t-Boc-N-amido-PEG4-alcohol
t-Boc-N-amido-PEG4-alcohol
Irritant
t-Boc-N-amido-PEG4-alcohol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
t-Boc-N-amido-PEG4-alcohol
Supplier:
Apollo Scientific
About this item
CAS Number:
106984-09-2
:
Formula:
C₁₃H₂₇NO₆
MDL Number:
MFCD07779195
:
293.36 g/mol
:
GHS07
Storage Temperature:
Freezer