Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PEG3-carboxylic acid tert-Butyl ester
PEG3-carboxylic acid tert-Butyl ester
PEG3-carboxylic acid tert-Butyl ester
:
Apollo Scientific
PEG3-carboxylic acid tert-Butyl ester
:
Apollo Scientific
:
133803-81-3
:
:
C₁₁H₂₂O₅
:
MFCD20926380
:
234.29 g/mol
:
GHS07
:
Ambient