Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nicotinamide
Nicotinamide 99%
Nicotinamide 99%
:
Apollo Scientific
Nicotinamide 99%
:
Apollo Scientific
:
98-92-0
:
:
C₆H₆N₂O
:
MFCD00006395
:
128…132 °C
:
1.4 g/cm³ (25 °C)
:
150 °C
:
122.13 g/mol
:
202-713-4
:
GHS07
:
Ambient