Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nigericin sodium salt
Nigericin sodium salt
Nigericin sodium salt
Toxic
Nigericin sodium salt
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
Nigericin sodium salt
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
28643-80-3
:
MFCD27977549
Formula:
C₄₀H₆₇NaO₁₁
MDL Number:
MFCD00036825
:
746.94 g/mol
Storage Temperature:
Refrigerator
UN:
3462
ADR:
6.1,III
:
GHS06
Related Information
Documentation
Certificates
Search by Lot Number
Documentation