Cesium chloride 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Boiling Pt:1303 °C
Melting Pt:645 °C
Density:3.99 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-600-2
:GHS07