
Okadaic acid free acid ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78111-17-8
:
Formula:C₄₄H₆₈O₁₃
MDL Number:MFCD00083455
:805.02 g/mol
Melting Pt:164…166 °C
Density:1.28 g/cm³ (25 °C)
Storage Temperature:Freezer
UN:3462
ADR:6.1,I
:GHS06