Order Entry
Germany
ContactUsLinkComponent
Antimycin A
Toxic
Antimycin A
Catalog # APOSBIM0142-50MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Antimycin A
Catalog # APOSBIM0142-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :548.63 g/mol
Melting Pt:149…150 °C
Storage Temperature:Freezer
UN:3462
ADR:6.1,II
 :GHS06
CAS Number:1397-94-0
Formula:C₂₈H₄₀N₂O₉
MDL Number:MFCD01779723