0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
DL-Homocysteinethiolactone hydrochloride
DL-Homocysteinethiolactone hydrochloride 97%
DL-Homocysteinethiolactone hydrochloride 97%
:
Apollo Scientific
DL-Homocysteinethiolactone hydrochloride 97%
:
Apollo Scientific
:
6038-19-3
:
:
C₄H₇NOS
:
MFCD00012724
:
153.62 g/mol
:
202…204 °C
:
227-923-3
:
GHS07
:
Ambient