Hypericin (from Hypericum perforatum)
Catalog # APOSBIH1002-10MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:504.45 g/mol
Melting Pt:320 °C (dec.)
EINECS:208-941-0
:GHS07
Storage Temperature:Freezer
CAS Number:548-04-9
Formula:C₃₀H₁₆O₈
MDL Number:MFCD00016683