0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
G418 disulphate
G418 disulphate
G418 disulphate
G418 disulphate
Supplier:
Apollo Scientific
Certificates
Expand image
G418 disulphate
Supplier:
Apollo Scientific
About this item
CAS Number:
108321-42-2
:
Formula:
C₂₀H₄₄N₄O₁₈S₂
MDL Number:
MFCD00058314
:
692.72 g/mol
:
GHS07
:
GHS08
Storage Temperature:
Refrigerator