Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluticasone propionate
Fluticasone propionate
Fluticasone propionate
Health
Fluticasone propionate
Supplier:
Apollo Scientific
Certificates
Expand image
Health
Fluticasone propionate
Supplier:
Apollo Scientific
About this item
CAS Number:
80474-14-2
:
Formula:
C₂₅H₃₁F₃O₅S
MDL Number:
MFCD00866007
:
500.57 g/mol
Melting Pt:
275 °C
:
GHS08
Storage Temperature:
Ambient