0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Phenyl-4,6-O-benzylidene-beta-D-glucopyranoside
Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98% min
Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98% min
Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98% min
Supplier:
Apollo Scientific
Certificates
Expand image
Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98% min
Supplier:
Apollo Scientific
About this item
CAS Number:
75829-66-2
:
Formula:
C₁₉H₂₀O₆
MDL Number:
MFCD00136366
:
344.363 g/mol
Storage Temperature:
Ambient