Order Entry
Germany
ContactUsLinkComponent
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside 98% min
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside 98% min
Catalog # APOSBICL4355-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside 98% min
Catalog # APOSBICL4355-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :282.29 g/mol
Storage Temperature:Ambient
CAS Number:3162-96-7
Formula:C₁₄H₁₈O₆
MDL Number:MFCD00006819