Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Curcumin
Curcumin 98%
Curcumin 98%
Curcumin 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Curcumin 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
458-37-7
:
Formula:
C₂₁H₂₀O₆
MDL Number:
MFCD00008365
:
368.39 g/mol
Melting Pt:
175 °C
EINECS:
207-280-5
:
GHS07
Storage Temperature:
Freezer
Related Information
Documentation
Certificates
Search by Lot Number
Documentation