Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Supplier:
Apollo Scientific
Certificates
Expand image
Coomassie brilliant blue G-250
Supplier:
Apollo Scientific
About this item
CAS Number:
6104-58-1
:
Formula:
C₄₇H₄₈N₃NaO₇S₂
MDL Number:
MFCD00078482
:
854.04 g/mol
EINECS:
228-058-4
:
GHS07
Storage Temperature:
Ambient