

Cytochalasin B
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14930-96-2
:
Formula:C₂₉H₃₇NO₅
MDL Number:MFCD00077704
:479.62 g/mol
Melting Pt:215…223 °C
Storage Temperature:Freezer
EINECS:239-000-2
UN:1544
ADR:6.1,II
:GHS06
:GHS08