Order Entry
Germany
ContactUsLinkComponent
Sodium pyruvate
Sodium pyruvate
Catalog # APOSBIB6270-500G
Supplier:  Apollo Scientific
CAS Number:  
Sodium pyruvate
Catalog # APOSBIB6270-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :110.04 g/mol
Melting Pt:>300 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586
Melting Pt:>300 °C