
3-Acetylpyridine adenine dinucleotide 92+% (oxidised form)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86-08-8
:
Formula:C₂₂H₂₈N₆O₁₄P₂
MDL Number:MFCD00078882
:662.44 g/mol
EINECS:201-649-4
:GHS07
Storage Temperature:Freezer