
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51963-61-2
:987-65-5
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MDL Number:MFCD00150754
:605.19 g/mol
Melting Pt:176 °C (dec.)
EINECS:213-579-1
:GHS07
Storage Temperature:Refrigerator