Order Entry
Germany
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Irritant
Supplier:   Apollo Scientific
Bromophenol blue
Supplier:  Apollo Scientific
Certificates
Bromophenol blue

2 Options Available Below

Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Melting Pt:279 °C
  • Storage Temperature:Ambient

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.