Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Biotin 99%

D-(+)-Biotin 99%

Irritant
Supplier:   Apollo Scientific
D-(+)-Biotin 99%
Supplier:  Apollo Scientific
Certificates
D-(+)-Biotin 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Melting Pt:231…233 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.