Order Entry
Germany
ContactUsLinkComponent
Zinc acetate dihydrate ACS, Reag. USP, Reag. Ph. Eur.
Zinc acetate dihydrate ACS, Reag. USP, Reag. Ph. Eur.
Catalog # APOSBIA1317-1KG
Supplier:  Apollo Scientific
CAS Number:  
Zinc acetate dihydrate ACS, Reag. USP, Reag. Ph. Eur.
Catalog # APOSBIA1317-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :219.51 g/mol
Melting Pt:237 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961