Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Agarose
Agarose molecular sieve grade
Agarose molecular sieve grade
Agarose molecular sieve grade
Supplier:
Apollo Scientific
Certificates
Expand image
Agarose molecular sieve grade
Supplier:
Apollo Scientific
About this item
CAS Number:
9012-36-6
:
MDL Number:
MFCD00081294
:
GHS07
Storage Temperature:
Ambient