Order Entry
Germany
ContactUsLinkComponent
L(+)-Asparagine monohydrate ≥99%
L(+)-Asparagine monohydrate ≥99%
Catalog # APOSBIA0725-250G
Supplier:  Apollo Scientific
CAS Number:  
L(+)-Asparagine monohydrate ≥99%
Catalog # APOSBIA0725-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :150.13 g/mol
Melting Pt:233…235 °C
Density:1.6 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:5794-13-8
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038