Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Alanine ethyl ester hydrochloride
L-Alanine ethyl ester hydrochloride
L-Alanine ethyl ester hydrochloride
APOSBIA0106-25G
:
Apollo Scientific
:
L-Alanine ethyl ester hydrochloride
APOSBIA0106-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
153.61 g/mol
:
78…80 °C
:
GHS07
:
Ambient
:
1115-59-9
:
C₅H₁₁NO₂
:
MFCD00063662