0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Ampicillin sodium salt
Ampicillin sodium salt
Ampicillin sodium salt
:
Apollo Scientific
Ampicillin sodium salt
:
Apollo Scientific
:
69-52-3
:
:
C₁₆H₁₈N₃NaO₄S
:
MFCD00064313
:
371.39 g/mol
:
215 °C (dec.)
:
200-708-1
:
GHS07
:
GHS08
:
Refrigerator