di-Sodium β-glycerophosphate pentahydrate 97+%
Catalog # APOSBI4838-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:306.11 g/mol
Melting Pt:102…104 °C
EINECS:212-464-3
:GHS07
Storage Temperature:Ambient
CAS Number:13408-09-8
:819-83-0
Formula:C₃H₇Na₂O₆P·5H₂O
MDL Number:MFCD00002135