Orders LinkContactUsLinkComponent
Grounding cables, b.safe
Grounding cables, b.safe
Supplier:   Bohlender
Grounding cables, b.safe
Supplier:  Bohlender
Certificates
Grounding cables, b.safe
Grounding cables, b.safe
11 Options Available Below
Product Details & Documents
For earth connection of vessels and components made of static dissipative materials.
  • Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
  • Customised versions can be offered on request
  • Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 590-0337
    Grounding cable with 2× clip
    Grounding cable with 2× clip
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,30 €
    33,30 €
    1 * 1 ST
       33,30 €
  • 590-0343
    Grounding cable with 2× ring cable lug M6
    Grounding cable with 2× ring cable lug M6
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,90 €
    25,90 €
    1 * 1 ST
       25,90 €
  • 590-0340
    Grounding cable with clip and banana plug
    Grounding cable with clip and banana plug
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • 590-0338
    Grounding cable with clip and ring cable lug M6
    Grounding cable with clip and ring cable lug M6
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,00 €
    35,00 €
    1 * 1 ST
       35,00 €
  • 590-0345
    Grounding cable with clip and stainless steel strap
    Grounding cable with clip and stainless steel strap
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,00 €
    62,00 €
    1 * 1 ST
       62,00 €
  • 590-0341
    Grounding cable with MC plug and ring cable lug M6
    Grounding cable with MC plug and ring cable lug M6
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,80 €
    25,80 €
    1 * 1 ST
       25,80 €
  • 590-0344
    Grounding cable with 2× ring cable lug M6
    Grounding cable with 2× ring cable lug M6
    3,0 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,80 €
    28,80 €
    1 * 1 ST
       28,80 €
  • 590-0342
    Grounding cable with clip and clip for tube Ø 32 mm
    Grounding cable with clip and clip for tube Ø 32 mm
    3,0 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,70 €
    32,70 €
    1 * 1 ST
       32,70 €
  • 590-0339
    Grounding cable with clip and ring cable lug M6
    Grounding cable with clip and ring cable lug M6
    3,0 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,90 €
    34,90 €
    1 * 1 ST
       34,90 €
  • BOHLR606-05
    Grounding cable with clip and 13 - 18 mm Ø tube clip
    Grounding cable with clip and 13 - 18 mm Ø tube clip
    3,0 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • BOHLR607-03
    Grounding cable with ring cable plug M6 and 32 mm Ø tube clip
    Grounding cable with ring cable plug M6 and 32 mm Ø tube clip
    3,0 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,90 €
    29,90 €
    1 * 1 ST
       29,90 €