Human recombinant siglec-10 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,01 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-2, Llama IgG2b Fc Tag, low endotoxin at 2 μg/ml (100 µl/well) can bind Anti-Human CD22 with a linear range of 4-63 ng/ml (QC tested).
- Gene ID:P20273-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:BL-CAM|SIGLEC-2|CD22|Siglec2|SIGLEC2|SIGLEC2FLJ22814
- Protein/peptide name:Siglec-2
- Purity:>95%
- Molecular weight:103,0 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT