Order Entry
Germany
ContactUsLinkComponent
 
Mouse recombinant IGF-I R (from HEK293)
 
Mouse recombinant IGF-I R (from HEK293)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse IGF-I R, His Tag (Cat. No. ACRBIGR-M5223) at 5 μg/ml (100 μl/well) can bind Human IGF-I, Fc Tag (Cat. No. ACRBIG1-H4269) with a linear range of 0,04 - 0,6 μg/ml (QC tested)
  • Gene ID:
    Q60751-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    MGC18216|JTK13|CD221|IGFR|MGC142170|IGF1R|MGC142172
  • Protein/peptide name:
    IGF-I R
  • Purity:
    >90%
  • Molecular weight:
    81,3 kDa, 24,3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

 

Frequently Bought Together