Marmoset recombinant OX40 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Marmoset
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Human OX40 Ligand, Fc Tag (Cat. No. ACRBOXL-H526x) at 5 μg/ml (100 μl/well) can bind Marmoset OX40, His Tag (Cat. No. ACRBOX0-M522b) with a linear range of 0,078 - 1,25 μg/ml (QC tested).; Measured by its binding ability in a functional ELISA. immobilised Marmoset OX40, His Tag (Cat. No. ACRBOX0-M522b) at 2 μg/ml (100 μl/well) can bind Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag (Cat. No. ACRBOXL-R5259) with a linear range of 0,2 - 3 ng/ml (Routinely tested)
- Gene ID:F7DHA3-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/peptide name:OX40
- Purity:>90%
- Molecular weight:21,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT