Human recombinant PD-1 (from HEK293 cells)
Catalog # MSPPPD1H5221-100UG
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,01 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) at 2 μg/mL (100 μL/well) can bind Human PD-L2, Mouse IgG1 Fc Tag (Cat. No. PD2-H52A5) with a linear range of 10-156 ng/mL (Routinely tested). Loaded Human PD-1, His Tag, low endotoxin (Cat. No. PD1-H522a) on HIS1K Biosensor, can bind Human PD-L1, Fc Tag (Cat. No. PD1-H5258) with an affinity constant of 23.3 nM as determined in BLI assay (ForteBio Octet Red96e) (QC tested). Loaded Human PD-1, His Tag, low endotoxin (Cat. No. PD1-H522a) on HIS1K Biosensor, can bind Human PD-L2, Fc Tag (HPLC-verified) (Cat. No. PD2-H5251) with an affinity constant of 15.7 nM as determined in BLI assay (ForteBio Octet Red96e) (QC tested). Loaded Human PD-L1, Fc Tag (Cat. No. PD1-H5258) on Protein A Biosensor, can bind Human Human PD-1, His Tag, low endotoxin (Cat. No. PD1-H522a) with an affinity constant of 4.8 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human PD-L2, Fc Tag (HPLC verified) (Cat. No. PD2-H5251) on Protein A Biosensor, can bind Human Human PD-1, His Tag, low endotoxin (Cat. No. PD1-H522a) with an affinity constant of 0.9 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q15116
- Protein synonyms:CD279|PDCD1|PD1|SLEB2
- Protein/peptide name:PD-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:16,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Human PD-1/PDCD1 Protein, His Tag, low endotoxin (MALS verified).