Order Entry
Germany
ContactUsLinkComponent
Mouse recombinant Glypican 2 (from HEK293 cells)
Mouse recombinant Glypican 2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant Glypican 2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Recombinant Human Midkine Protein, CF at 2 μg/ml (100 μl/well) can bind Mouse Glypican 2, His Tag (ACRBGP2-M52H5) with a linear range of 2 - 13 ng/ml (QC tested)
  • Gene ID:
    Q8BKV1-1
  • Protein synonyms:
    GPC2|Glypican 2
  • Protein/peptide name:
    Glypican 2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    60,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications