Order Entry
Germany
ContactUsLinkComponent
Human recombinant insulin R (from HEK293 cells)
Human recombinant insulin R (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant insulin R (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (SPR verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Human Insulin R (28-944), His Tag at 5 μg/ml (100 μl/well) can bind Human IGF-II, Fc Tag with a linear range of 0,078 to 1,25 μg/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human Insulin R (28-944), His Tag at 5 μg/ml (100 μl/well) can bind Insulin Biotin Conjugate with a linear range of 0,02 to 0,313 μg/ml (Routinely tested). Human Insulin R (28-944), His Tag captured on CM5 chip via anti-His antibody, can bind Human Insulin with an affinity constant of 249 nm as determined in a SPR assay (Biacore T200) (QC tested).
  • Gene ID:
    P06213-2
  • Protein synonyms:
    INSR|Insulin receptor|IR|CD220
  • Protein/peptide name:
    Insulin R
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    106,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications