Order Entry
Germany
ContactUsLinkComponent
Human recombinant latent TGF-beta 1 (from HEK293)
Human recombinant latent TGF-beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant latent TGF-beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised human ITGAV and ITGB6 heterodimer protein, His Tag and Tag Free (Cat. No. ACRBIT6-H52E1) at 5 μg/ml (100 μl/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. ACRBTG1-H82Qb) with a linear range of 0,5 - 4 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human ITGAV and ITGB8 heterodimer protein, His Tag and Tag Free (Cat. No. ACRBIT8-H52W4) at 5 μg/ml (100 μl/well) can bind biotinylated human latent TGF-beta 1, His,Avitag (Cat. No. ACRBTG1-H82Qb) with a linear range of 0,2 - 4 ng/ml (routinely tested).
  • Gene ID:
    P01137-1 (C33S)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
  • Protein/peptide name:
    Latent TGF-beta 1
  • Purity:
    >90%
  • Molecular weight:
    45,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications