Human recombinant integrin alpha V beta 5 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human vitronectin at 5 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB5 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT5-H82Wa) with a linear range of 0,01 to 0,156 μg/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised human fibronectin at 5 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB5 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT5-H82Wa) with a linear range of 0,156 - 5 μg/ml (routinely tested).
- Gene ID:P06756-1 (ITGAV) and P18084-1 (ITGB5)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Integrin alpha V beta 5|ITGAV and ITGB5
- Protein/peptide name:Integrin alpha V beta 5
- Purity:>95%
- Molecular weight:114,7 kDa (ITGAV) and 81,9 kDa (ITGB5)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT